Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014630.4 | NP_001014630.3 | 20 - 21 | 6880 | 76 | |
NM_001260232.2 | NP_001247161.2 | 20 - 21 | 6880 | 76 | ||
NM_001260235.2 | NP_001247164.2 | 20 - 21 | 5950 | 76 | ||
NM_001260237.2 | NP_001247166.2 | 20 - 21 | 6880 | 76 | ||
NM_001260240.2 | NP_001247169.2 | 20 - 21 | 8800 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |