Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142429.3 | NP_650686.1 | 2 - 3 | 1902 | 75 | |
GenBank mRNA | AF244916.1 | - | 2 - 3 | 1870 | 75 | |
AY118662.1 | - | 2 - 3 | 1870 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |