Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRNexo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142458.4 | NP_650715.3 | 3 - 4 | 305 | 74 | |
GenBank mRNA | EF680280.1 | - | 3 - 4 | 238 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |