Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260260.1 | NP_001247189.1 | 4 - 5 | 443 | 71 | |
NM_001260261.1 | NP_001247190.1 | 4 - 5 | 676 | 71 | ||
NM_142509.5 | NP_650766.1 | 3 - 4 | 468 | 71 | ||
GenBank mRNA | AY051929.1 | - | 3 - 4 | 444 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |