Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obp93a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142688.1 | NP_650945.1 | 3 - 4 | 393 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |