Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142736.3 | NP_650993.1 | 1 | 371 | 76 | |
GenBank mRNA | AY071724.1 | - | 1 | 372 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |