Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142879.3 | NP_651136.1 | 2 - 3 | 1543 | 64 | |
GenBank mRNA | AY118904.1 | - | 2 - 3 | 1512 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |