Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142961.5 | NP_651218.1 | 2 - 3 | 1025 | 90 | |
GenBank mRNA | AJ242992.1 | - | 2 - 3 | 1000 | 90 | |
BT150181.1 | - | 2 - 3 | 1023 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |