Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142961.5 | NP_651218.1 | 4 - 5 | 1804 | 66 | |
GenBank mRNA | AJ242992.1 | - | 4 - 5 | 1779 | 66 | |
BT150181.1 | - | 4 - 5 | 1802 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |