Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043289.2 | NP_001036754.1 | 4 - 5 | 5250 | 75 | |
NM_001043290.2 | NP_001036755.2 | 4 - 5 | 5247 | 75 | ||
NM_001260360.1 | NP_001247289.1 | 4 - 5 | 5250 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |