Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ets96B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143029.3 | NP_651286.3 | 6 - 7 | 1711 | 82 | |
NM_206567.2 | NP_996290.1 | 5 - 6 | 1910 | 82 | ||
GenBank mRNA | AY119230.1 | - | 5 - 6 | 2027 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |