Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170250.1 | NP_001163721.1 | 3 - 4 | 1849 | 66 | |
NM_143084.2 | NP_651341.2 | 4 - 5 | 1837 | 66 | ||
GenBank mRNA | BT030842.1 | - | 3 - 4 | 1787 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |