Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bili | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170251.2 | NP_001163722.1 | 10 - 11 | 2430 | 68 | |
NM_001170252.2 | NP_001163723.1 | 10 - 11 | 2419 | 68 | ||
NM_001170253.2 | NP_001163724.1 | 10 - 11 | 2514 | 68 | ||
NM_001170254.2 | NP_001163725.1 | 10 - 11 | 2450 | 68 | ||
NM_001170255.1 | NP_001163726.1 | 10 - 11 | 2570 | 68 | ||
NM_001300592.1 | NP_001287521.1 | 10 - 11 | 2823 | 68 | ||
NM_143085.3 | NP_651342.3 | 10 - 11 | 2527 | 68 | ||
GenBank mRNA | AY094737.1 | - | 10 - 11 | 2489 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |