Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: danr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143086.3 | NP_651343.1 | 1 | 1646 | 160 | |
GenBank mRNA | AY084187.1 | - | 1 | 1641 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |