Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ymp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300601.1 | NP_001287530.1 | 4 - 5 | 440 | 68 | |
NM_001300602.1 | NP_001287531.1 | 4 - 5 | 440 | 68 | ||
NM_143147.3 | NP_651404.1 | 4 - 5 | 440 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |