Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpr97Ea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276077.1 | NP_001263006.1 | 2 - 3 | 448 | 77 | |
NM_143272.2 | NP_651529.2 | 2 - 3 | 460 | 77 | ||
GenBank mRNA | BT150156.1 | - | 2 - 3 | 460 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |