Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr98a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143307.2 | NP_651564.1 | 2 - 3 | 1326 | 92 | |
GenBank mRNA | BT050571.1 | - | 2 - 3 | 1150 | 92 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Drosophila melanogaster |