Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Moca-cyp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170287.3 | NP_001163758.1 | 1 - 2 | 411 | 67 | |
NM_143361.6 | NP_651618.3 | 1 - 2 | 411 | 67 | ||
NM_170367.3 | NP_733246.1 | 1 - 2 | 411 | 67 | ||
GenBank mRNA | AY051595.1 | - | 1 - 2 | 392 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |