Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Moca-cyp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170287.3 | NP_001163758.1 | 4 - 5 | 1535 | 75 | |
NM_143361.6 | NP_651618.3 | 5 - 6 | 1368 | 75 | ||
NM_170367.3 | NP_733246.1 | 5 - 6 | 810 | 75 | ||
GenBank mRNA | AY051595.1 | - | 5 - 6 | 1349 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |