Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dgt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143414.4 | NP_651671.1 | 3 - 4 | 1901 | 124 | |
GenBank mRNA | AY069433.1 | - | 3 - 4 | 1858 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |