Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cisd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143427.4 | NP_651684.1 | 2 - 3 | 414 | 63 | |
GenBank mRNA | AY071722.1 | - | 2 - 3 | 414 | 63 | |
AY095084.1 | - | 2 - 3 | 413 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |