Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap-D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143452.3 | NP_651709.1 | 2 - 3 | 683 | 69 | |
GenBank mRNA | AY122183.1 | - | 2 - 3 | 658 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |