Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jdp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143550.3 | NP_651807.1 | 2 - 3 | 188 | 63 | |
NM_170501.2 | NP_733380.1 | 2 - 3 | 188 | 63 | ||
NM_170502.2 | NP_733381.1 | 2 - 3 | 188 | 63 | ||
GenBank mRNA | AF192462.1 | - | 2 - 3 | 162 | 63 | |
AF192463.1 | - | 2 - 3 | 162 | 63 | ||
AY060331.1 | - | 2 - 3 | 161 | 63 | ||
BT011341.1 | - | 2 - 3 | 191 | 63 | ||
BT132968.1 | - | 2 - 3 | 187 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |