Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PH4alphaNE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143557.2 | NP_651814.1 | 5 - 6 | 1254 | 77 | |
GenBank mRNA | AF495538.1 | - | 5 - 6 | 1254 | 77 | |
AY051566.1 | - | 5 - 6 | 1254 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |