Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143603.2 | NP_651860.2 | 3 - 4 | 775 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |