Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276223.2 | NP_001263152.1 | 3 - 4 | 706 | 93 | |
NM_143637.5 | NP_651894.3 | 3 - 4 | 706 | 93 | ||
GenBank mRNA | AY060677.1 | - | 3 - 4 | 687 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |