Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eip93F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104395.2 | NP_001097865.1 | 4 - 5 | 919 | 65 | |
NM_001275888.1 | NP_001262817.1 | 3 - 4 | 765 | 65 | ||
NM_143745.3 | NP_652002.2 | 3 - 4 | 765 | 65 | ||
GenBank mRNA | BT010034.1 | - | 4 - 5 | 919 | 65 | |
BT031093.1 | - | 2 - 3 | 683 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |