Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt302C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144369.3 | NP_652626.1 | 1 - 2 | 400 | 78 | |
GenBank mRNA | AY060891.1 | - | 1 - 2 | 376 | 78 | |
BT006007.1 | - | 1 - 2 | 398 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |