Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CG46339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104469.3 | NP_001097939.1 | 6 - 7 | 1667 | 90 | |
NM_001260394.2 | NP_001247323.1 | 6 - 7 | 1370 | 90 | ||
NM_001276072.1 | NP_001263001.1 | 7 - 8 | 1703 | 90 | ||
NM_001363935.1 | NP_001350863.1 | 4 - 5 | 1238 | 90 | ||
NM_001363936.1 | NP_001350864.1 | 5 - 6 | 1571 | 90 | ||
NM_144487.3 | NP_652744.2 | 5 - 6 | 1334 | 90 | ||
GenBank mRNA | AF480087.1 | - | 5 - 6 | 1311 | 90 | |
AY122083.1 | - | 5 - 6 | 1374 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster |