Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or82a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_164323.1 | NP_730794.1 | 3 - 4 | 848 | 77 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |