Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rheb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169068.3 | NP_730950.2 | 3 - 4 | 631 | 112 | |
NM_169069.3 | NP_730951.2 | 4 - 5 | 465 | 112 | ||
GenBank mRNA | AY094697.1 | - | 4 - 5 | 427 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |