Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144554.3 | NP_001138026.1 | 2 - 3 | 740 | 77 | |
NM_001275437.2 | NP_001262366.1 | 2 - 3 | 740 | 77 | ||
NM_169206.3 | NP_731211.1 | 2 - 3 | 740 | 77 | ||
GenBank mRNA | AY071369.1 | - | 2 - 3 | 738 | 77 | |
D50542.1 | - | 2 - 3 | 543 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |