Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SelR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169369.3 | NP_731523.1 | 3 - 4 | 650 | 99 | |
NM_169370.4 | NP_731524.1 | 3 - 4 | 535 | 99 | ||
GenBank mRNA | BT011487.1 | - | 3 - 4 | 436 | 99 | |
KX531816.1 | - | 2 - 3 | 275 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |