Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300368.1 | NP_001287297.1 | 1 - 2 | 1389 | 60 | |
NM_169451.2 | NP_731670.1 | 2 - 3 | 1395 | 60 | ||
GenBank mRNA | BT133437.1 | - | 2 - 3 | 1393 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |