Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080168.3 | NP_524907.2 | 3 - 4 | 2029 | 66 | |
NM_169531.2 | NP_731828.1 | 2 - 3 | 1928 | 66 | ||
NM_169532.2 | NP_731829.1 | 4 - 5 | 2262 | 66 | ||
NM_169533.2 | NP_731830.2 | 3 - 4 | 2029 | 66 | ||
GenBank mRNA | AY058617.1 | - | 3 - 4 | 2016 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |