Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cv-c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104316.2 | NP_001097786.1 | 9 - 10 | 5355 | 77 | |
NM_001104318.2 | NP_001097788.1 | 10 - 11 | 4688 | 77 | ||
NM_169583.3 | NP_731907.1 | 2 - 3 | 535 | 77 | ||
GenBank mRNA | BT044146.1 | - | 2 - 3 | 527 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |