Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stumps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104322.2 | NP_001097792.1 | 3 - 4 | 715 | 59 | |
NM_169588.2 | NP_731923.2 | 2 - 3 | 633 | 59 | ||
GenBank mRNA | BT024930.1 | - | 3 - 4 | 713 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |