Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btsz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014622.3 | NP_001014622.3 | 5 - 6 | 3532 | 71 | |
NM_001014625.3 | NP_001014625.1 | 3 - 4 | 2626 | 71 | ||
NM_001144584.3 | NP_001138056.1 | 3 - 4 | 2626 | 71 | ||
NM_001260180.2 | NP_001247109.1 | 4 - 5 | 2959 | 71 | ||
NM_001275653.1 | NP_001262582.1 | 5 - 6 | 3559 | 71 | ||
GenBank mRNA | AY229969.1 | - | 3 - 4 | 2610 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |