Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169637.4 | NP_732005.2 | 9 - 10 | 876 | 70 | |
GenBank mRNA | BT133132.1 | - | 9 - 10 | 828 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |