Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: msps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170156.2 | NP_001163627.1 | 10 - 11 | 5655 | 119 | |
NM_001260200.2 | NP_001247129.1 | 11 - 12 | 5751 | 119 | ||
NM_001260201.2 | NP_001247130.1 | 11 - 12 | 5751 | 119 | ||
NM_001260202.1 | NP_001247131.1 | 10 - 11 | 5318 | 119 | ||
NM_169698.3 | NP_732105.2 | 10 - 11 | 5655 | 119 | ||
GenBank mRNA | BT023496.1 | - | 10 - 11 | 5632 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |