Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: msps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170156.2 | NP_001163627.1 | 14 - 15 | 6522 | 60 | |
NM_001260200.2 | NP_001247129.1 | 15 - 16 | 6618 | 60 | ||
NM_001260201.2 | NP_001247130.1 | 15 - 16 | 6594 | 60 | ||
NM_001260202.1 | NP_001247131.1 | 14 - 15 | 6185 | 60 | ||
NM_169698.3 | NP_732105.2 | 14 - 15 | 6498 | 60 | ||
GenBank mRNA | BT023496.1 | - | 14 - 15 | 6475 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |