Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mhcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275694.1 | NP_001262623.1 | 2 - 3 | 1681 | 101 | |
NM_169703.1 | NP_732111.1 | 1 - 2 | 1744 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |