Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275745.1 | NP_001262674.1 | 1 | 983 | 121 | |
NM_001300447.1 | NP_001287376.1 | 1 | 983 | 121 | ||
NM_169763.3 | NP_732244.3 | 1 | 983 | 121 | ||
GenBank mRNA | BT030410.1 | - | 1 | 984 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |