Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tinc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275757.1 | NP_001262686.1 | 8 - 9 | 4431 | 87 | |
NM_169780.5 | NP_732278.2 | 9 - 10 | 4463 | 87 | ||
NM_176509.4 | NP_788686.1 | 9 - 10 | 4518 | 87 | ||
NM_176510.5 | NP_788687.3 | 9 - 10 | 4428 | 87 | ||
NM_176511.3 | NP_788688.2 | 9 - 10 | 4476 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |