Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gukh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032021.2 | NP_001027192.1 | 3 - 4 | 1788 | 96 | |
NM_001104364.2 | NP_001097834.2 | 3 - 4 | 1788 | 96 | ||
NM_169845.2 | NP_732393.1 | 3 - 4 | 1788 | 96 | ||
NM_169846.2 | NP_732394.1 | 3 - 4 | 1788 | 96 | ||
GenBank mRNA | BT023826.1 | - | 3 - 4 | 1785 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |