Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GluRIIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043268.2 | NP_001036733.1 | 4 - 5 | 1014 | 70 | |
GenBank mRNA | BT056325.1 | - | 4 - 5 | 1012 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |