Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Takl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169924.3 | NP_732554.1 | 2 - 3 | 380 | 77 | |
GenBank mRNA | BT022955.1 | - | 2 - 3 | 349 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |