Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNF4Agamma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043271.3 | NP_001036736.2 | 3 - 4 | 427 | 75 | |
NM_001104384.3 | NP_001097854.1 | 3 - 4 | 397 | 75 | ||
NM_001104385.2 | NP_001097855.1 | 3 - 4 | 733 | 75 | ||
NM_001170200.2 | NP_001163671.1 | 7 - 8 | 2919 | 75 | ||
NM_001170201.2 | NP_001163672.1 | 7 - 8 | 2801 | 75 | ||
NM_080509.3 | NP_536757.2 | 7 - 8 | 1652 | 75 | ||
NM_169946.2 | NP_732594.1 | 7 - 8 | 1823 | 75 | ||
NM_169947.3 | NP_732598.1 | 7 - 8 | 2978 | 75 | ||
GenBank mRNA | AY070541.1 | - | 7 - 8 | 2955 | 75 | |
AY166752.1 | - | 7 - 8 | 2962 | 75 | ||
BT003626.1 | - | 1652 | 75 | |||
BT010233.1 | - | 7 - 8 | 1821 | 75 | ||
BT029676.1 | - | 3 - 4 | 427 | 75 | ||
BT029715.1 | - | 3 - 4 | 397 | 75 | ||
BT120301.1 | - | 7 - 8 | 1671 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |