Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104392.3 | NP_001097862.1 | 6 - 7 | 2123 | 84 | |
NM_001104393.3 | NP_001097863.1 | 6 - 7 | 692 | 84 | ||
NM_169970.3 | NP_732661.1 | 5 - 6 | 2218 | 84 | ||
NM_169971.3 | NP_732662.2 | 6 - 7 | 2123 | 84 | ||
GenBank mRNA | BT044550.1 | - | 6 - 7 | 2083 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |