Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: loco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260311.1 | NP_001247240.1 | 3 - 4 | 2730 | 56 | |
NM_001260312.2 | NP_001247241.1 | 3 - 4 | 2239 | 56 | ||
NM_170030.2 | NP_732773.1 | 4 - 5 | 4436 | 56 | ||
NM_170032.3 | NP_732775.1 | 4 - 5 | 3099 | 56 | ||
NM_170033.3 | NP_732776.1 | 3 - 4 | 2266 | 56 | ||
GenBank mRNA | AY118939.1 | - | 4 - 5 | 4409 | 56 | |
BT122165.1 | - | 3 - 4 | 2730 | 56 | ||
BT133319.1 | - | 3 - 4 | 2245 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |